ENA292596033 Enamine

{6,8-dichloroimidazo[1,2-a]pyridin-2-yl}methanol

  • MDL number MFCD16040116
  • Cas Number : 1235441-30-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62041

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62041
Availability details for
Quantity
  • Availability
  • Add to Cart