ENA292596038 Enamine

N-[6-(4-bromophenoxy)pyridin-3-yl]-2-chloroacetamide

  • MDL number MFCD16040121
  • Cas Number : 1235438-88-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-62077

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62077
Availability details for
Quantity
  • Availability
  • Add to Cart