ENA292596042 Enamine

(1-methyl-1H-pyrazol-4-yl)methyl acetate

  • MDL number MFCD16040125
  • Cas Number : 1235441-11-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62093

Safety Information

msds https://www.enaminestore.com/catalog/EN300-62093
Availability details for
Quantity
  • Availability
  • Add to Cart