ENA292596045 Enamine

3-amino-4-(morpholin-4-yl)-1lambda6-thiolane-1,1-dione dihydrochloride

  • MDL number MFCD16040128
  • Cas Number : 1235439-96-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-62112

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62112
Availability details for
Quantity
  • Availability
  • Add to Cart