ENA292596046 Enamine

2-amino-2-methylbutanamide hydrochloride

  • MDL number MFCD16040129
  • Cas Number : 18305-22-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 235...237
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62116

Safety Information

msds https://www.enaminestore.com/catalog/EN300-62116
Availability details for
Quantity
  • Availability
  • Add to Cart