ENA292596050 Enamine

2-[4-(cyclopentyloxy)phenyl]ethan-1-amine hydrochloride

  • MDL number MFCD16040133
  • Cas Number : 1235440-38-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62162

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62162
Availability details for
Quantity
  • Availability
  • Add to Cart