ENA292596051 Enamine

[3-(cyclopentyloxy)phenyl]methanamine hydrochloride

  • MDL number MFCD16040134
  • Cas Number : 1235441-67-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 206...208
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62163

Safety Information

msds https://www.enaminestore.com/catalog/EN300-62163
Availability details for
Quantity
  • Availability
  • Add to Cart