ENA292596052 Enamine

2-amino-N-cyclopropyl-3-phenylpropanamide hydrochloride

  • MDL number MFCD16040135
  • Cas Number : 1251922-95-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 145...147
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62165

Safety Information

msds https://www.enaminestore.com/catalog/EN300-62165
Availability details for
Quantity
  • Availability
  • Add to Cart