ENA292596055 Enamine

5-[2-(4-fluorophenyl)ethenyl]-3-methyl-1,2-oxazol-4-amine

  • MDL number MFCD16040139
  • Cas Number : 2135539-69-2
  • Molecular Weight : 218.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-62217

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62217
Availability details for
Quantity
  • Availability
  • Add to Cart