ENA292596059 Enamine

1-[2-(morpholin-4-yl)ethyl]piperidin-4-amine trihydrochloride

  • MDL number MFCD16040143
  • Cas Number : 1235439-73-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 306...308
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62275

Safety Information

msds https://www.enaminestore.com/catalog/EN300-62275
Availability details for
Quantity
  • Availability
  • Add to Cart