ENA292596062 Enamine

1-(propan-2-yl)-2-sulfanyl-1,4-dihydropyrimidin-4-one

  • MDL number MFCD16040146
  • Cas Number : 1235439-08-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62279

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62279
Availability details for
Quantity
  • Availability
  • Add to Cart