ENA292596067 Enamine

methyl 2-(1,4-diazepan-1-yl)pyridine-4-carboxylate hydrochloride

  • MDL number MFCD16040153
  • Cas Number : 1235439-20-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62337

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62337
Availability details for
Quantity
  • Availability
  • Add to Cart