ENA292596068 Enamine

3-[3-(3-fluorophenyl)-1,2,4-oxadiazol-5-yl]piperidine hydrochloride

  • MDL number MFCD16040154
  • Cas Number : 1235441-45-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-62364

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62364
Availability details for
Quantity
  • Availability
  • Add to Cart