ENA292596070 Enamine

N-(2-methylphenyl)-1,3-thiazolidine-4-carboxamide hydrochloride

  • MDL number MFCD16040156
  • Cas Number : 1251923-39-3
  • Molecular Weight : 258.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62368

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62368
Availability details for
Quantity
  • Availability
  • Add to Cart