ENA292596074 Enamine

N-(2-aminoethyl)-2-cyclohexylacetamide hydrochloride

  • MDL number MFCD16040160
  • Cas Number : 1235440-90-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-62386

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62386
Availability details for
Quantity
  • Availability
  • Add to Cart