ENA292596078 Enamine

6-phenyl-3,4-dihydro-2H-1-benzopyran-4-amine hydrochloride

  • MDL number MFCD16040166
  • Cas Number : 191608-31-8
  • Molecular Weight : 261.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62429

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62429
Availability details for
Quantity
  • Availability
  • Add to Cart