ENA292596081 Enamine

4-chloro-2-fluoro-6-(trifluoromethyl)aniline

  • MDL number MFCD16040170
  • Cas Number : 1235441-41-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62547

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62547
Availability details for
Quantity
  • Availability
  • Add to Cart