ENA292596132 Enamine

N'-hydroxy-4-(4-methylpiperidin-1-yl)butanimidamide

  • MDL number MFCD09936637
  • Cas Number : 1016797-22-0
  • Molecular Weight : 199.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-60279

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60279
Availability details for
Quantity
  • Availability
  • Add to Cart