ENA292596133 Enamine

6-hydroxy-3-(3-methoxyphenyl)-2-sulfanyl-3,4-dihydropyrimidin-4-one

  • MDL number MFCD05739458
  • Cas Number : 1147192-75-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-61588

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61588
Availability details for
Quantity
  • Availability
  • Add to Cart