ENA292596138 Enamine

N-methyl-1-(pyridin-2-yl)piperidin-4-amine

  • MDL number MFCD12048853
  • Cas Number : 405058-06-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-60370

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60370
Availability details for
Quantity
  • Availability
  • Add to Cart