ENA292596144 Enamine

2-(6-methyl-1H-indol-3-yl)acetonitrile

  • MDL number MFCD11048378
  • Cas Number : 500777-89-9
  • Molecular Weight : 170.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-60373

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60373
Availability details for
Quantity
  • Availability
  • Add to Cart