ENA292596145 Enamine

5-methylpiperidin-2-one

  • MDL number MFCD13178700
  • Cas Number : 3298-16-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 55-56
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-61604

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61604
Availability details for
Quantity
  • Availability
  • Add to Cart