ENA292596151 Enamine

3-methyl-5H,6H,7H-indeno[5,6-b]furan-2-carboxylic acid

  • MDL number MFCD11104765
  • Cas Number : 1152583-23-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61624

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61624
Availability details for
Quantity
  • Availability
  • Add to Cart