ENA292596153 Enamine

3-{[(1-methyl-1H-imidazol-2-yl)sulfanyl]methyl}benzoic acid

  • MDL number MFCD09941376
  • Cas Number : 1017046-67-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61626

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61626
Availability details for
Quantity
  • Availability
  • Add to Cart