ENA292596154 Enamine

2-(6-chloro-1H-indol-3-yl)acetonitrile

  • MDL number MFCD03428598
  • Cas Number : 61220-58-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 99...101
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60376

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-60376
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart