ENA292596158 Enamine

4-[(2-methoxyethane)sulfonyl]benzoic acid

  • MDL number MFCD09942739
  • Cas Number : 1016888-12-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160...162
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60483

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60483
Availability details for
Quantity
  • Availability
  • Add to Cart