ENA292596159 Enamine

4-(chloromethyl)-2-(furan-2-yl)-5-methyl-1,3-oxazole

  • MDL number MFCD08056337
  • Cas Number : 141399-54-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-61633

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61633
Availability details for
Quantity
  • Availability
  • Add to Cart