ENA292596163 Enamine

[1-(3-nitropyridin-2-yl)piperidin-4-yl]methanol

  • MDL number MFCD13242900
  • Cas Number : 1235440-20-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61638

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61638
Availability details for
Quantity
  • Availability
  • Add to Cart