ENA292596165 Enamine

3-amino-2-methylbutan-2-ol

  • MDL number MFCD16040002
  • Cas Number : 6291-17-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60610

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-60610
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart