ENA292596167 Enamine

N-(4-fluorophenyl)-2-(N'-hydroxycarbamimidoyl)acetamide

  • Cas Number : 219528-73-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61643

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61643
Availability details for
Quantity
  • Availability
  • Add to Cart