ENA292596172 Enamine

N-[(4-fluorophenyl)methyl]-1,3-benzothiazol-2-amine

  • MDL number MFCD05792693
  • Cas Number : 923893-82-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61649

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61649
Availability details for
Quantity
  • Availability
  • Add to Cart