ENA292596173 Enamine

2-[3-(4-methylphenyl)propanamido]acetic acid

  • MDL number MFCD11648372
  • Cas Number : 1098367-46-4
  • Molecular Weight : 221.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61650

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61650
Availability details for
Quantity
  • Availability
  • Add to Cart