ENA292596184 Enamine

1-(2-methoxyphenyl)cyclopropane-1-carboxylic acid

  • MDL number MFCD00297169
  • Cas Number : 74205-24-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 131...133
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61674

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-61674
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart