ENA292596186 Enamine

5-nitro-1H-indole-3-carboxylic acid

  • MDL number MFCD06203492
  • Cas Number : 6958-37-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61676

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61676
Availability details for
Quantity
  • Availability
  • Add to Cart