ENA292596188 Enamine

2-hydroxy-1-(1,2,3,4-tetrahydroquinolin-1-yl)ethan-1-one

  • MDL number MFCD08442900
  • Cas Number : 73251-26-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 80...82
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61678

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61678
Availability details for
Quantity
  • Availability
  • Add to Cart