ENA292596191 Enamine

6-hydroxy-3-(2-methylphenyl)-2-sulfanyl-3,4-dihydropyrimidin-4-one

  • MDL number MFCD00815747
  • Cas Number : 93341-41-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 206...208
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61685

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61685
Availability details for
Quantity
  • Availability
  • Add to Cart