ENA292596192 Enamine

3-propoxyphenol

  • MDL number MFCD11181933
  • Cas Number : 16533-50-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-61688

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61688
Availability details for
Quantity
  • Availability
  • Add to Cart