ENA292596197 Enamine

2-phenyl-3,4-dihydropyrimidin-4-one

  • MDL number MFCD00234618
  • Cas Number : 33643-94-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 204...206
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61706

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61706
Availability details for
Quantity
  • Availability
  • Add to Cart