ENA292596198 Enamine

5-(2-bromo-5-fluorophenyl)-2H-1,2,3,4-tetrazole

  • MDL number MFCD04035678
  • Cas Number : 870562-41-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61707

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61707
Availability details for
Quantity
  • Availability
  • Add to Cart