ENA292596202 Enamine

3-(pyrimidin-2-yl)-4,5-dihydro-1,2,4-oxadiazol-5-one

  • MDL number MFCD12815757
  • Cas Number : 1184003-44-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61712

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61712
Availability details for
Quantity
  • Availability
  • Add to Cart