ENA292596205 Enamine

3-(pyridin-4-yl)-4,5-dihydro-1,2,4-oxadiazol-5-one

  • MDL number MFCD12774152
  • Cas Number : 102227-52-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 253-255
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61715

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61715
Availability details for
Quantity
  • Availability
  • Add to Cart