ENA292596211 Enamine

3-methyl-[1,2,4]triazolo[4,3-a]pyridine-6-carboxylic acid

  • MDL number MFCD07376944
  • Cas Number : 1031619-88-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 307...309
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60836

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-60836
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart