ENA292596213 Enamine

[1,2,4]triazolo[4,3-a]pyridine-6-carboxylic acid

  • MDL number MFCD07376943
  • Cas Number : 933708-92-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-60837

Safety Information

signalword Warning
pcodes P264,P280,P305+P351+P338,P337+P313
hcodes Xi
MSDS https://www.enaminestore.com/catalog/EN300-60837
pictograms GHS07
hstatements H319
Availability details for
Quantity
  • Availability
  • Add to Cart