ENA292596218 Enamine

2-(pyridin-3-yloxy)pyridine-3-carboxylic acid

  • MDL number MFCD11102531
  • Cas Number : 214758-27-7
  • Molecular Weight : 216.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-61730

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61730
Availability details for
Quantity
  • Availability
  • Add to Cart