ENA292596239 Enamine

N-ethyl-3-hydroxybenzamide

  • MDL number MFCD08729290
  • Cas Number : 15788-98-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160...162
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61770

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61770
Availability details for
Quantity
  • Availability
  • Add to Cart