ENA292596241 Enamine

1-(2-methylphenyl)cyclohexane-1-carboxylic acid

  • MDL number MFCD11037175
  • Cas Number : 1039931-32-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-61772

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61772
Availability details for
Quantity
  • Availability
  • Add to Cart