ENA292596242 Enamine

6-bromo-7-fluoro-1,2,3,4-tetrahydroquinolin-2-one

  • MDL number MFCD12174779
  • Cas Number : 1156389-00-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 210...212
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61773

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61773
Availability details for
Quantity
  • Availability
  • Add to Cart