ENA292596248 Enamine

2-amino-2-(2,4-difluorophenyl)acetonitrile

  • MDL number MFCD09041124
  • Cas Number : 189138-13-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-61779

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61779
Availability details for
Quantity
  • Availability
  • Add to Cart