ENA292596251 Enamine

methyl 4-[(2,4-dioxo-1,3-thiazolidin-3-yl)methyl]benzoate

  • MDL number MFCD03948191
  • Cas Number : 749216-29-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-61782

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61782
Availability details for
Quantity
  • Availability
  • Add to Cart