ENA292596252 Enamine

2-[(piperazin-1-yl)methyl]-1H-1,3-benzodiazole dihydrochloride

  • MDL number MFCD10023086
  • Cas Number : 1177362-39-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61785

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61785
Availability details for
Quantity
  • Availability
  • Add to Cart